Assay Details
| Gene Symbol: Jam3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023277.4 | NP_075766.1 | 3 - 4 | 308 | 114 | |
| GenBank mRNA | AJ300304.1 | - | 3 - 4 | 268 | 114 | |
| AK003326.1 | - | 3 - 4 | 268 | 114 | ||
| AK008187.1 | - | 3 - 4 | 301 | 114 | ||
| AK013156.1 | - | 3 - 4 | 267 | 114 | ||
| AK032833.1 | - | 3 - 4 | 268 | 114 | ||
| AK160648.1 | - | 3 - 4 | 306 | 114 | ||
| BC024357.1 | - | 3 - 4 | 280 | 4973185 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jam3
Genomic Map