Assay Details
Gene Symbol: Kri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145416.3 | NP_663391.2 | 15 - 16 | 1506 | 111 | |
GenBank mRNA | BC021438.1 | - | 15 - 16 | 1496 | 5039543 | 111 |
CX562209.1 | - | 5 - 6 | 533 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
