Assay Details
| Gene Symbol: Icam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008319.2 | NP_032345.2 | 8 - 9 | 2086 | 73 | |
| GenBank mRNA | BC113140.1 | - | 8 - 9 | 2034 | 40048010 | 73 |
| U06483.1 | - | 8 - 9 | 2034 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Icam5
Genomic Map