Assay Details
Gene Symbol: Raver1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027911.3 | NP_082187.1 | 11 - 12 | 2030 | 68 | |
GenBank mRNA | AK051444.1 | - | 11 - 12 | 2043 | 68 | |
AK079191.1 | - | 11 - 12 | 2041 | 68 | ||
AK084478.1 | - | 11 - 12 | 2042 | 68 | ||
AK138596.1 | - | 11 - 12 | 2041 | 68 | ||
AK168436.1 | - | 11 - 12 | 2052 | 68 | ||
BC025589.1 | - | 3 - 4 | 365 | 68 | ||
BC043698.1 | - | 3 - 4 | 465 | 68 | ||
BC049920.1 | - | 2 - 3 | 211 | 68 | ||
BC058271.1 | - | 11 - 12 | 2030 | 6490833 | 68 | |
BC108392.1 | - | 11 - 12 | 2029 | 6393833 | 68 |
Gene Symbol: Gm38431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038081.2 | 11 - 12 | 2050 | 68 | ||
GenBank mRNA | AK004919.1 | - | 11 - 12 | 2043 | 68 | |
AY275472.1 | - | 11 - 12 | 2029 | 68 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
