Assay Details
| Gene Symbol: Fxyd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111073.2 | NP_001104543.1 | 5 - 6 | 767 | 78 | |
| NM_001287213.1 | NP_001274142.1 | 6 - 7 | 639 | 78 | ||
| NM_001287217.1 | NP_001274146.1 | 6 - 7 | 630 | 78 | ||
| NM_008761.4 | NP_032787.1 | 6 - 7 | 627 | 78 | ||
| GenBank mRNA | AK003068.1 | - | 6 - 7 | 410 | 78 | |
| AK155763.1 | - | 5 - 6 | 747 | 78 | ||
| AK169966.1 | - | 6 - 7 | 410 | 78 | ||
| AK170810.1 | - | 6 - 7 | 415 | 78 | ||
| BC013340.1 | - | 6 - 7 | 412 | 78 | ||
| BC031112.1 | - | 6 - 7 | 399 | 78 | ||
| U72680.1 | - | 6 - 7 | 433 | 78 | ||
More Information
Set Membership: |
|

Fxyd5
Genomic Map