Assay Details
| Gene Symbol: Mag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010758.2 | NP_034888.1 | 6 - 7 | 1123 | 60 | |
| GenBank mRNA | BC101944.1 | - | 6 - 7 | 1096 | 40039200 | 60 |
| BC103521.1 | - | 6 - 7 | 1096 | 40039204 | 60 | |
| BC103522.1 | - | 6 - 7 | 1096 | 60 | ||
| BC103523.1 | - | 7 - 8 | 970 | 60 | ||
| M31811.1 | - | 6 - 7 | 1123 | 60 | ||
| X07849.1 | - | 1 - 2 | 46 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mag
Genomic Map