| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900005579.mm.1 |
NM_001253883.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005579.mm.1 |
NM_001253883.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005579.mm.1 |
NM_011099.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005579.mm.1 |
NM_011099.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005579.mm.1 |
TC0900000751.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900005578.mm.1 |
NM_001253883.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005578.mm.1 |
NM_001253883.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005578.mm.1 |
NM_011099.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005578.mm.1 |
NM_011099.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005578.mm.1 |
TC0900000751.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900005575.mm.1 |
NM_001253883.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005575.mm.1 |
NM_001253883.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005575.mm.1 |
NM_011099.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005575.mm.1 |
NM_011099.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005575.mm.1 |
TC0900000751.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900005580.mm.1 |
NM_001253883.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005580.mm.1 |
NM_001253883.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005580.mm.1 |
NM_011099.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005580.mm.1 |
NM_011099.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900005580.mm.1 |
TC0900000751.mm.1 |
MTA TC |