Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900002081.mm.1 |
NM_001115130.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002081.mm.1 |
NM_001115130.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002081.mm.1 |
NM_172765.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002081.mm.1 |
NM_172765.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002081.mm.1 |
TC0900000273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900002080.mm.1 |
NM_001115130.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002080.mm.1 |
NM_001115130.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002080.mm.1 |
NM_172765.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002080.mm.1 |
NM_172765.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002080.mm.1 |
TC0900000273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900002082.mm.1 |
NM_001115130.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002082.mm.1 |
NM_001115130.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002082.mm.1 |
NM_172765.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002082.mm.1 |
NM_172765.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002082.mm.1 |
TC0900000273.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900002077.mm.1 |
NM_001115130.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002077.mm.1 |
NM_001115130.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002077.mm.1 |
NM_172765.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002077.mm.1 |
NM_172765.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002077.mm.1 |
TC0900000273.mm.1 |
MTA TC |