Assay Details
Gene Symbol: Psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026545.3 | NP_080821.3 | 2 - 3 | 469 | 62 | |
GenBank mRNA | AK002921.1 | - | 2 - 3 | 308 | 62 | |
AK003436.1 | - | 2 - 3 | 310 | 62 | ||
AK020132.1 | - | 2 - 3 | 328 | 62 | ||
AK027973.1 | - | 2 - 3 | 273 | 62 | ||
AK051257.1 | - | 2 - 3 | 321 | 62 | ||
AK150493.1 | - | 2 - 3 | 312 | 62 | ||
AK150982.1 | - | 2 - 3 | 276 | 62 | ||
AK153329.1 | - | 2 - 3 | 310 | 62 | ||
AK159216.1 | - | 2 - 3 | 278 | 62 | ||
AK159795.1 | - | 2 - 3 | 294 | 62 | ||
AK166718.1 | - | 2 - 3 | 303 | 62 | ||
AK168886.1 | - | 2 - 3 | 277 | 62 | ||
BC004075.1 | - | 2 - 3 | 287 | 3591796 | 62 | |
BC005717.1 | - | 2 - 3 | 197 | 2609781 | 62 | |
CF732286.1 | - | 2 - 3 | 469 | 62 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK013113.1 | - | Chr. - - [-] on Build | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK153329.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
