| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900017245.mm.1 |
NM_001199556.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900017245.mm.1 |
NM_001199556.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900017245.mm.1 |
NM_178737.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900017245.mm.1 |
NM_178737.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900017245.mm.1 |
TC0900002083.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900017243.mm.1 |
NM_001199556.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900017243.mm.1 |
NM_001199556.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900017243.mm.1 |
NM_178737.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900017243.mm.1 |
NM_178737.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900017243.mm.1 |
TC0900002083.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900017254.mm.1 |
TC0900002083.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900017253.mm.1 |
NM_178737.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900017253.mm.1 |
NM_178737.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900017253.mm.1 |
TC0900002083.mm.1 |
MTA TC |