Assay Details
| Gene Symbol: Capns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009795.3 | NP_033925.2 | 1 - 2 | 42 | 73 | |
| GenBank mRNA | BC018352.1 | - | 1 - 2 | 36 | 3156021 | 73 |
| BC090988.1 | - | 1 - 2 | 42 | 30246016 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC018352.1, BC090988.1, NM_009795.3 transcripts.
Set Membership: |
|

Capns1
Genomic Map