Assay Details
| Gene Symbol: Capns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009795.3 | NP_033925.2 | 4 - 5 | 388 | 93 | |
| GenBank mRNA | AF058298.1 | - | 3 - 4 | 367 | 93 | |
| AK028092.1 | - | 2 - 3 | 134 | 93 | ||
| AK132819.1 | - | 3 - 4 | 198 | 93 | ||
| BC018352.1 | - | 4 - 5 | 382 | 3156021 | 93 | |
| BC090988.1 | - | 4 - 5 | 388 | 30246016 | 93 | |
More Information
Set Membership: |
|

Capns1
Genomic Map