Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700023457.mm.1 |
TC0700002685.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700023457.mm.1 |
NM_011874.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023457.mm.1 |
NM_011874.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023455.mm.1 |
TC0700002685.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700023455.mm.1 |
NM_011874.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023455.mm.1 |
NM_011874.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023454.mm.1 |
TC0700002685.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700023454.mm.1 |
NM_011874.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023454.mm.1 |
NM_011874.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023460.mm.1 |
TC0700002685.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700023460.mm.1 |
NM_011874.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700023460.mm.1 |
NM_011874.2 |
Refseq |