Assay Details
| Gene Symbol: Psmc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011874.2 | NP_036004.2 | 3 - 4 | 392 | 66 | |
| GenBank mRNA | AK050677.1 | - | 3 - 4 | 370 | 66 | |
| AK077507.1 | - | 3 - 4 | 355 | 66 | ||
| AK150989.1 | - | 3 - 4 | 347 | 66 | ||
| AK153991.1 | - | 3 - 4 | 375 | 66 | ||
| AK160644.1 | - | 3 - 4 | 354 | 66 | ||
| AK160718.1 | - | 3 - 4 | 346 | 66 | ||
| AK167041.1 | - | 3 - 4 | 337 | 66 | ||
| AK167528.1 | - | 3 - 4 | 347 | 66 | ||
| AK169224.1 | - | 3 - 4 | 355 | 66 | ||
| BC012708.1 | - | 3 - 4 | 353 | 4211677 | 66 | |
| BC092265.1 | - | 3 - 4 | 336 | 3671250 | 66 | |
| DV646128.1 | - | 3 - 4 | 402 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmc4
Genomic Map