Assay Details
Gene Symbol: Sertad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018820.4 | NP_061290.1 | 1 - 2 | 125 | 65 | |
GenBank mRNA | AF366401.1 | - | 1 - 2 | 168 | 65 | |
AK002324.1 | - | 1 - 2 | 126 | 65 | ||
AK004022.1 | - | 1 - 2 | 125 | 65 | ||
AK008303.1 | - | 1 - 2 | 126 | 65 | ||
BC016077.1 | - | 1 - 2 | 85 | 4486318 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF366401.1, AK002324.1, AK004022.1, AK008303.1, BC016077.1, NM_018820.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017513 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017512 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017511 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017518 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017515 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017517 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436878 | AF366401.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436878 | AK002324.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436878 | AK004022.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436878 | BC016077.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436878 | NM_018820.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436878 | AF366401.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436878 | AK002324.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436878 | AK004022.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436878 | BC016077.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P436878 | NM_018820.4 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 60777 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139573 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146216 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436878 | AF366401.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436878 | AK002324.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436878 | AK004022.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436878 | BC016077.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436878 | NM_018820.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436878 | AF366401.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436878 | AK002324.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436878 | AK004022.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436878 | BC016077.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P436878 | NM_018820.4 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P217211 | AF366401.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P217211 | AK002324.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P217211 | AK004022.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P217211 | AK008303.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P217211 | BC016077.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436878 | AF366401.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436878 | AK002324.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436878 | AK004022.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436878 | BC016077.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436878 | NM_018820.4 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436878 | AF366401.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436878 | AK002324.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436878 | AK004022.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436878 | BC016077.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P436878 | NM_018820.4 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017520 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017512 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017497 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017523 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017506 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017496 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017509 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017498 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017508 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017510 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017519 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017503 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017516 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017513 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017499 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017530 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017515 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017504 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017518 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017528 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017501 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017522 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017525 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25017527 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146216 | - | - |
Whole Mouse Genome G4122A | A_52_P217211 | AF366401.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P217211 | AK002324.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P217211 | AK004022.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P217211 | AK008303.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P217211 | BC016077.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P436878 | AF366401.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P436878 | AK002324.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P436878 | AK004022.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P436878 | BC016077.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P436878 | NM_018820.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P436878 | AF366401.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P436878 | AK002324.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P436878 | AK004022.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P436878 | BC016077.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P436878 | NM_018820.4 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108456 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102106 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146216 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06177809 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06631426 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06177801 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06177803 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017512 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017518 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017511 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017517 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017513 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25017515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25017552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017484 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017481 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017483 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06517300 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017504 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017503 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017512 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017506 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017537 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017477 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017490 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017502 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017535 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017478 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017534 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017475 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06177805 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017498 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017500 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06177815 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06080917 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06273891 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017489 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017517 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017485 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06440822 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06080913 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06440824 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017508 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017509 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06148614 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017518 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017525 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017487 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017526 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06080910 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06631426 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017530 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017476 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25017499 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06455701 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10551362 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551361 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551360 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551364 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551363 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417406_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10551359 | - | - |
Mouse Genome 430 2.0 Array | 1417406_at | - | - |
Mouse Genome 430 2.0 Array | 1417406_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 55942_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 55942_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700003655.mm.1 | TC0700000460.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700003655.mm.1 | NM_018820.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700003655.mm.1 | NM_018820.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700003657.mm.1 | TC0700000460.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700003657.mm.1 | NM_018820.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700003657.mm.1 | NM_018820.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700003658.mm.1 | TC0700000460.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700003658.mm.1 | NM_018820.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700003658.mm.1 | NM_018820.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700003654.mm.1 | TC0700000460.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700003654.mm.1 | NM_018820.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700003654.mm.1 | NM_018820.4 | Refseq |
Mouse Expression Set 430 | 1417406_PM_at | - | - |
Mouse Expression Set 430 | 1417406_at | - | - |
Murine Genome U74v2 Set | 160834_PM_at | - | - |
Murine Genome U74v2 Set | 160834_at | - | - |
Mouse Genome 430A 2.0 Array | 1417406_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417406_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417406_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417406_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 921416 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2680332 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2680332 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002680332 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2729162 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2729162 | - | - |
mouse-6 v1.1 (Illumina) | scl32896.3_171-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2680332 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl32896.3_171-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2680332 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2729162 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2729162 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32896.3_171-S | - | - |
