Assay Details
| Gene Symbol: Dmkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166173.1 | NP_001159645.1 | 18 - 19 | 1697 | 55 | |
| NM_001166174.1 | NP_001159646.1 | 16 - 17 | 1604 | 55 | ||
| NM_028618.2 | NP_082894.1 | 5 - 6 | 288 | 55 | ||
| NM_172899.4 | NP_766487.3 | 17 - 18 | 1649 | 55 | ||
| GenBank mRNA | AK003695.1 | - | 5 - 6 | 291 | 55 | |
| AK081753.1 | - | 18 - 19 | 1699 | 55 | ||
| AY444557.1 | - | 16 - 17 | 1561 | 55 | ||
| AY444558.1 | - | 5 - 6 | 291 | 55 | ||
| AY622962.1 | - | 5 - 6 | 269 | 55 | ||
| AY622963.1 | - | 16 - 17 | 1508 | 55 | ||
| BC064724.1 | - | 17 - 18 | 1642 | 30435974 | 55 | |
More Information
Set Membership: |
|

Dmkn
Genomic Map