Assay Details
Gene Symbol: Etv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007959.2 | NP_031985.2 | 1 - 2 | 94 | 81 | |
GenBank mRNA | AB221612.1 | - | 1 - 2 | 77 | 81 | |
L10427.1 | - | 1 - 2 | 94 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
