Assay Details
Gene Symbol: Syne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153577.2 | NP_705805.1 | 5 - 6 | 999 | 101 | |
GenBank mRNA | BC004761.1 | - | 5 - 6 | 948 | 3155758 | 101 |
BC023803.1 | - | 5 - 6 | 986 | 5352953 | 101 | |
BC056649.1 | - | 5 - 6 | 938 | 5036575 | 101 | |
BG063039.2 | - | 3 - 4 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P147803 | BC023803.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P147803 | BG063039.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P147803 | NM_153577.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P147803 | AK002612.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P147803 | BC023803.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P147803 | BG063039.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P147803 | NM_153577.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P147803 | AK002612.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 114934 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 613715 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 660166 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79938 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145965 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P147803 | BC023803.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P147803 | BG063039.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P147803 | NM_153577.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P147803 | AK002612.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P147803 | BC023803.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P147803 | BG063039.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P147803 | NM_153577.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P147803 | AK002612.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030946 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030940 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030933 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030931 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030943 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030951 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030934 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030937 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030935 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030942 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030949 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030938 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030944 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030936 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25030952 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06154250 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145965 | - | - |
Whole Mouse Genome G4122A | A_52_P147803 | BC023803.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P147803 | BG063039.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P147803 | NM_153577.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P147803 | AK002612.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P147803 | BC023803.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P147803 | BG063039.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P147803 | NM_153577.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P147803 | AK002612.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127678 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145965 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06582227 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06582224 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06582229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25031019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030953 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030972 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030961 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030965 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030952 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06464103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030943 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06466446 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06464102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030942 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030964 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06390431 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06027567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06582228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06464106 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06020328 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030955 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06000439 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030940 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030951 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06582224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030934 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030969 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030959 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030960 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030970 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030946 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030938 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25030944 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10551876 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551880 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551877 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551879 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551873 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551874 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551878 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551875 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427176_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427175_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10551872 | - | - |
Mouse Genome 430 2.0 Array | 1427176_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1427175_at | - | - |
Mouse Genome 430 2.0 Array | 1427176_s_at | - | - |
Mouse Genome 430 2.0 Array | 1427175_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233066_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233066_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700004844.mm.1 | TC0700000559.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700004844.mm.1 | NM_153577.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700004844.mm.1 | NM_153577.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700004856.mm.1 | TC0700000559.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700004856.mm.1 | NM_153577.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700004856.mm.1 | NM_153577.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700004843.mm.1 | TC0700000559.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700004843.mm.1 | NM_153577.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700004843.mm.1 | NM_153577.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700004852.mm.1 | TC0700000559.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700004852.mm.1 | NM_153577.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700004852.mm.1 | NM_153577.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700004855.mm.1 | TC0700000559.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700004855.mm.1 | NM_153577.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700004855.mm.1 | NM_153577.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700004857.mm.1 | TC0700000559.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700004857.mm.1 | NM_153577.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700004857.mm.1 | NM_153577.2 | Refseq |
Mouse Expression Set 430 | 1427175_PM_at | - | - |
Mouse Expression Set 430 | 1427175_at | - | - |
Mouse Expression Set 430 | 1427176_PM_s_at | - | - |
Mouse Expression Set 430 | 1427176_s_at | - | - |
Murine Genome U74v2 Set | 161008_at | - | - |
Murine Genome U74v2 Set | 161857_PM_r_at | - | - |
Murine Genome U74v2 Set | 161008_PM_at | - | - |
Murine Genome U74v2 Set | 161857_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1427176_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1427175_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427175_at | - | - |
Mouse Genome 430A 2.0 Array | 1427176_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427176_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427175_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427176_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427175_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2650433 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2650433 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002650433 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2619639 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2619639 | - | - |
mouse-6 v1.1 (Illumina) | scl32818.7.1_108-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2650433 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl32818.7.1_108-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650433 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2619639 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2619639 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32818.7.1_108-S | - | - |
