Assay Details
| Gene Symbol: 9530077C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026739.1 | NP_081015.1 | 1 - 2 | 169 | 65 | |
| GenBank mRNA | AK021129.1 | - | 1 - 2 | 205 | 65 | |
| AK160343.1 | - | 1 - 2 | 218 | 65 | ||
| AK220359.1 | - | 1 - 2 | 233 | 65 | ||
| BC054761.1 | - | 1 - 2 | 169 | 6413638 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK021129.1, BC054761.1, NM_026739.1 transcripts.
Set Membership: |
|

9530077C05Rik
Genomic Map