Assay Details
| Gene Symbol: Zfp810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145612.2 | NP_663587.1 | 1 - 2 | 88 | 129 | |
| GenBank mRNA | AK032556.1 | - | 1 - 2 | 128 | 129 | |
| BC005471.1 | - | 1 - 2 | 88 | 3496918 | 129 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK032556.1, BC005471.1, NM_145612.2 transcripts.
Set Membership: |
|

Zfp810
Genomic Map