Assay Details
Gene Symbol: Zfp810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145612.2 | NP_663587.1 | 6 - 7 | 600 | 79 | |
GenBank mRNA | AK032556.1 | - | 6 - 7 | 640 | 79 | |
BC005471.1 | - | 6 - 7 | 600 | 3496918 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26351263 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26351264 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26351261 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328883 | BC005471.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328883 | NM_145612.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328883 | BC005471.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P328883 | NM_145612.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281596 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545607 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164217 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478836 | AK040423.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478836 | AK080059.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478836 | AK131687.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478836 | AK165009.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478836 | AK165900.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478836 | BC030352.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478836 | BC038704.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478836 | NM_009229.4 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478836 | U00678.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 421770 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830380 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680373 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 764390 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628651 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160497 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 242224 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 751486 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 929295 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934590 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 209461 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 882969 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432460 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 30361 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 102698 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 159829 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 813734 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 70710 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 42893 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141000 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162863 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141541 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328883 | BC005471.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328883 | NM_145612.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328883 | BC005471.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P328883 | NM_145612.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328883 | BC005471.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328883 | NM_145612.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328883 | BC005471.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P328883 | NM_145612.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351265 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351249 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351278 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351260 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351273 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351285 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351252 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351271 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351248 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351258 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351262 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351269 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351281 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351274 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351282 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351266 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351275 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351263 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351259 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351264 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351284 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351255 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26351276 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141541 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162863 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06720871 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06720884 | - | - |
Whole Mouse Genome G4122A | A_51_P328883 | BC005471.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328883 | NM_145612.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328883 | BC005471.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P328883 | NM_145612.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140142 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131286 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141541 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162863 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07761244 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07761226 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07761231 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07761243 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26351261 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26351264 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26351263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26351078 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351258 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351251 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351244 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351266 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351297 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351241 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351293 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351236 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351238 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351269 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07842777 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351267 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351237 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351287 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351285 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351276 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351296 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351255 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351264 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351260 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351271 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351281 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351257 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07477698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351248 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07842776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351284 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07886948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351253 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07761224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351265 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351273 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08118484 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07824713 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351259 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08036243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26351240 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10591766 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591767 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591770 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591768 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591772 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591771 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591764 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591769 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10591765 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451566_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10591763 | - | - |
Mouse Genome 430 2.0 Array | 1447383_at | - | - |
Mouse Genome 430 2.0 Array | 1451566_at | - | - |
Mouse Genome 430 2.0 Array | 1451566_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1447383_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 235050_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 235050_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900016052.mm.1 | NM_145612.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900016052.mm.1 | NM_145612.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900016052.mm.1 | TC0900001905.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900016051.mm.1 | NM_145612.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900016051.mm.1 | NM_145612.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900016051.mm.1 | TC0900001905.mm.1 | MTA TC |
Affymetrix HT Mouse Genome 430A Array | 1451566_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451566_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 555808 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 50398 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000050398 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231164 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1231164 | - | - |
mouse-6 v1.1 (Illumina) | scl36114.9_96-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 50398 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl36114.9_96-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 50398 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1231164 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231164 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36114.9_96-S | - | - |
