Assay Details
| Gene Symbol: Rgl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023622.4 | NP_076111.2 | 12 - 13 | 1410 | 83 | |
| GenBank mRNA | AF237669.1 | - | 12 - 13 | 1369 | 83 | |
| AF239661.1 | - | 12 - 13 | 1411 | 83 | ||
| AK053550.1 | - | 12 - 13 | 1397 | 83 | ||
| AK131140.1 | - | 916 | 83 | |||
| AK134654.1 | - | 1392 | 83 | |||
More Information
Set Membership: |
|

Rgl3
Genomic Map