Assay Details
| Gene Symbol: Zfp61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009561.2 | NP_033587.2 | 4 - 5 | 669 | 100 | |
| GenBank mRNA | AK148158.1 | - | 4 - 5 | 487 | 100 | |
| AK149960.1 | - | 4 - 5 | 464 | 100 | ||
| BC006618.1 | - | 4 - 5 | 708 | 3967527 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK148158.1, AK149960.1, BC006618.1, NM_009561.2 transcripts.
Set Membership: |
|

Zfp61
Genomic Map