Assay Details
| Gene Symbol: Arpp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142655.1 | NP_001136127.1 | 2 - 3 | 158 | 92 | |
| NM_021548.4 | NP_067523.1 | 1 - 2 | 227 | 92 | ||
| GenBank mRNA | AK049760.1 | - | 1 - 2 | 49 | 92 | |
| BC040206.1 | - | 1 - 2 | 227 | 3485973 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049760.1, BC040206.1, NM_021548.4 transcripts.
Set Membership: |
|

Arpp19
Genomic Map