Assay Details
| Gene Symbol: Chek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007691.5 | NP_031717.2 | 10 - 11 | 1219 | 97 | |
| GenBank mRNA | AK011258.1 | - | 10 - 11 | 1163 | 97 | |
| AK033179.1 | - | 10 - 11 | 1189 | 97 | ||
| AK045336.1 | - | 10 - 11 | 1161 | 97 | ||
| AK166675.1 | - | 10 - 11 | 1220 | 97 | ||
| BC037613.1 | - | 9 - 10 | 932 | 97 | ||
| BC053394.1 | - | 10 - 11 | 1156 | 97 | ||
| BC064445.1 | - | 9 - 10 | 1618 | 97 | ||
| BC100386.1 | - | 9 - 10 | 1294 | 30481390 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chek1
Genomic Map