Assay Details
| Gene Symbol: Vsig2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020518.2 | NP_065264.2 | 2 - 3 | 342 | 80 | |
| GenBank mRNA | AK008920.1 | - | 2 - 3 | 304 | 80 | |
| BC099457.1 | - | 2 - 3 | 306 | 30789633 | 80 | |
More Information
Set Membership: |
|

Vsig2
Genomic Map