Assay Details
| Gene Symbol: Acrv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007391.2 | NP_031417.2 | 1 - 2 | 141 | 144 | |
| GenBank mRNA | AK005705.1 | - | 1 - 2 | 141 | 144 | |
| AK030129.1 | - | 1 - 2 | 107 | 144 | ||
| BC100459.1 | - | 1 - 2 | 141 | 144 | ||
| BC139101.1 | - | 1 - 2 | 100 | 8862123 | 144 | |
| BC139103.1 | - | 1 - 2 | 100 | 8862125 | 144 | |
| U31992.1 | - | 1 - 2 | 112 | 144 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005705.1, BC139101.1, BC139103.1, NM_007391.2, U31992.1 transcripts.
Set Membership: |
|

Acrv1
Genomic Map