Assay Details
| Gene Symbol: Acrv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007391.2 | NP_031417.2 | 3 - 4 | 758 | 66 | |
| GenBank mRNA | AK005705.1 | - | 3 - 4 | 758 | 66 | |
| BC100459.1 | - | 3 - 4 | 757 | 66 | ||
| BC139101.1 | - | 3 - 4 | 717 | 8862123 | 66 | |
| BC139103.1 | - | 3 - 4 | 717 | 8862125 | 66 | |
| U31992.1 | - | 3 - 4 | 729 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263667 | U31992.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263667 | BC100459.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263667 | BC139101.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263667 | BC139103.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263667 | NM_007391.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263667 | U31992.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263667 | BC100459.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263667 | BC139101.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263667 | BC139103.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P263667 | NM_007391.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 599371 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410922 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868829 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 167586 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145514 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263667 | U31992.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263667 | BC100459.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263667 | BC139101.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263667 | BC139103.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263667 | NM_007391.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263667 | U31992.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263667 | BC100459.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263667 | BC139101.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263667 | BC139103.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P263667 | NM_007391.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26426309 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01905984 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145514 | - | - |
| Whole Mouse Genome G4122A | A_51_P263667 | U31992.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263667 | BC100459.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263667 | BC139101.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263667 | BC139103.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263667 | NM_007391.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263667 | U31992.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263667 | BC100459.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263667 | BC139101.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263667 | BC139103.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P263667 | NM_007391.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126195 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145514 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07754460 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07754474 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07754468 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07754481 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07754456 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07754470 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07754484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26426275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07561164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07866892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07801569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07858883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07872538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07754483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07754482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07858886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07866890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07866886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07693902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07866885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07501920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07754467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26426281 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10584256 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584255 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584258 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584257 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418878_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10584254 | - | - |
| Mouse Genome 430 2.0 Array | 1418878_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418878_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11451_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11451_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900002585.mm.1 | NM_007391.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900002585.mm.1 | NM_007391.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900002585.mm.1 | TC0900000348.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900002586.mm.1 | NM_007391.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900002586.mm.1 | NM_007391.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900002586.mm.1 | TC0900000348.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418878_at | - | - |
| Mouse Expression Set 430 | 1418878_PM_at | - | - |
| Murine Genome U74v2 Set | 161403_PM_r_at | - | - |
| Murine Genome U74v2 Set | 92897_PM_at | - | - |
| Murine Genome U74v2 Set | 92897_at | - | - |
| Murine Genome U74v2 Set | 161403_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418878_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418878_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418878_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418878_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4070161 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004070161 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2677129 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2677129 | - | - |
| mouse-6 v1.1 (Illumina) | scl37125.3.1_81-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4070161 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37125.3.1_81-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4070161 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2834622 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2834622 | - | - |

Acrv1
Genomic Map