Assay Details
| Gene Symbol: Tmprss5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030709.2 | NP_109634.2 | 3 - 4 | 290 | 76 | |
| GenBank mRNA | AB016229.1 | - | 3 - 4 | 298 | 76 | |
| AB016230.1 | - | 3 - 4 | 290 | 76 | ||
| AB041037.1 | - | 3 - 4 | 247 | 76 | ||
| AK029714.1 | - | 4 - 5 | 1137 | 76 | ||
| BC132151.1 | - | 2 - 3 | 236 | 40130428 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB016229.1 transcripts.
Set Membership: |
|

Tmprss5
Genomic Map