Assay Details
| Gene Symbol: Tmprss5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030709.2 | NP_109634.2 | 6 - 7 | 668 | 60 | |
| GenBank mRNA | AK029714.1 | - | 7 - 8 | 1515 | 60 | |
| BC132151.1 | - | 5 - 6 | 614 | 40130428 | 60 | |
| BC145027.1 | - | 5 - 6 | 596 | 9053575 | 60 | |
More Information
Set Membership: |
|

Tmprss5
Genomic Map