Assay Details
| Gene Symbol: Tmprss5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030709.2 | NP_109634.2 | 9 - 10 | 1046 | 79 | |
| GenBank mRNA | AK029714.1 | - | 10 - 11 | 1893 | 79 | |
| BC132151.1 | - | 8 - 9 | 992 | 40130428 | 79 | |
| BC145027.1 | - | 8 - 9 | 974 | 9053575 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss5
Genomic Map