Assay Details
| Gene Symbol: Mpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025837.2 | NP_080113.1 | 4 - 5 | 521 | 67 | |
| GenBank mRNA | AF244360.1 | - | 4 - 5 | 503 | 67 | |
| AK003289.1 | - | 2 - 3 | 281 | 67 | ||
| AK003896.1 | - | 4 - 5 | 521 | 67 | ||
| AK132777.1 | - | 4 - 5 | 522 | 67 | ||
| BC009068.1 | - | 4 - 5 | 499 | 2650791 | 67 | |
| BC051163.1 | - | 4 - 5 | 499 | 6466145 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpi
Genomic Map