Assay Details
| Gene Symbol: Ptpn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019651.2 | NP_062625.2 | 3 - 4 | 930 | 114 | |
| GenBank mRNA | AF013490.1 | - | 3 - 4 | 400 | 114 | |
| BC053017.1 | - | 3 - 4 | 930 | 5708923 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn9
Genomic Map