Assay Details
| Gene Symbol: Csk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013057.1 | - | 2 - 3 | 180 | 64 | |
| AK136876.1 | - | 10 - 11 | 1606 | 64 | ||
| AK137297.1 | - | 10 - 11 | 1019 | 64 | ||
| AK156058.1 | - | 10 - 11 | 1157 | 64 | ||
| AK170815.1 | - | 10 - 11 | 1173 | 64 | ||
| BC018394.1 | - | 10 - 11 | 1132 | 4206879 | 64 | |
| BC052006.1 | - | 10 - 11 | 1190 | 5685297 | 64 | |
| BC095962.1 | - | 12 - 13 | 1513 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Csk
Genomic Map