Assay Details
| Gene Symbol: Csk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK137297.1 | - | 7 - 8 | 757 | 76 | |
| AK156058.1 | - | 7 - 8 | 895 | 76 | ||
| AK170815.1 | - | 7 - 8 | 911 | 76 | ||
| BC018394.1 | - | 7 - 8 | 870 | 4206879 | 76 | |
| BC052006.1 | - | 7 - 8 | 928 | 5685297 | 76 | |
| BC095962.1 | - | 9 - 10 | 1251 | 76 | ||
| U05247.1 | - | 5 - 6 | 622 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Csk
Genomic Map