Assay Details
| Gene Symbol: Pinlyp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037143.2 | NP_001032220.1 | 1 - 2 | 83 | 67 | |
| GenBank mRNA | AK009589.1 | - | 1 - 2 | 88 | 67 | |
| AK009604.1 | - | 1 - 2 | 85 | 67 | ||
| AK009778.1 | - | 1 - 2 | 83 | 67 | ||
| AK009924.1 | - | 1 - 2 | 85 | 67 | ||
| BC139039.1 | - | 1 - 2 | 149 | 8862061 | 67 | |
| BC145686.1 | - | 1 - 2 | 149 | 40130989 | 67 | |
More Information
Set Membership: |
|

Pinlyp
Genomic Map