Assay Details
| Gene Symbol: Lypd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182785.3 | NP_877586.1 | 2 - 3 | 348 | 67 | |
| GenBank mRNA | AK077071.1 | - | 2 - 3 | 349 | 67 | |
| BC049744.1 | - | 1 - 2 | 148 | 6772266 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lypd4
Genomic Map