Assay Details
| Gene Symbol: Syde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027875.1 | NP_082151.1 | 3 - 4 | 1152 | 64 | |
| GenBank mRNA | AK004654.1 | - | 3 - 4 | 1134 | 64 | |
| AK197128.1 | - | 1 - 2 | 48 | 64 | ||
| AK198535.1 | - | 1 - 2 | 48 | 64 | ||
| AK204625.1 | - | 1 - 2 | 48 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syde1
Genomic Map