Assay Details
| Gene Symbol: Pdxk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172134.2 | NP_742146.1 | 3 - 4 | 344 | 75 | |
| GenBank mRNA | AK039194.1 | - | 3 - 4 | 326 | 75 | |
| AK080846.1 | - | 3 - 4 | 320 | 75 | ||
| AK145470.1 | - | 3 - 4 | 308 | 75 | ||
| AK166078.1 | - | 3 - 4 | 327 | 75 | ||
| AK166464.1 | - | 3 - 4 | 309 | 75 | ||
| BC027745.1 | - | 3 - 4 | 381 | 5064695 | 75 | |
| BC032171.1 | - | 3 - 4 | 315 | 75 | ||
| BI143426.1 | - | 3 - 4 | 373 | 75 | ||
More Information
Set Membership: |
|

Pdxk
Genomic Map