Assay Details
| Gene Symbol: Icosl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015790.3 | NP_056605.1 | 1 - 2 | 166 | 67 | |
| GenBank mRNA | AK041320.1 | - | 1 - 2 | 176 | 67 | |
| AK041421.1 | - | 1 - 2 | 176 | 67 | ||
| BC029227.1 | - | 1 - 2 | 166 | 4217333 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041421.1, BC029227.1, NM_015790.3 transcripts.
Set Membership: |
|

Icosl
Genomic Map