Assay Details
| Gene Symbol: Icosl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015790.3 | NP_056605.1 | 3 - 4 | 641 | 80 | |
| GenBank mRNA | AF199027.1 | - | 3 - 4 | 550 | 80 | |
| AF216747.1 | - | 2 - 3 | 484 | 80 | ||
| AF394451.1 | - | 484 | 80 | |||
| AK041320.1 | - | 2 - 3 | 1109 | 80 | ||
| AK041421.1 | - | 3 - 4 | 651 | 80 | ||
| BC029227.1 | - | 3 - 4 | 641 | 4217333 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Icosl
Genomic Map