Assay Details
Gene Symbol: Casp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009809.5 | NP_033939.1 | 6 - 7 | 886 | 90 | |
GenBank mRNA | AF092997.1 | - | 5 - 6 | 731 | 90 | |
AJ007750.1 | - | 6 - 7 | 751 | 90 | ||
AK081645.1 | - | 6 - 7 | 839 | 90 | ||
BC101959.1 | - | 6 - 7 | 824 | 40044873 | 90 | |
BC101960.1 | - | 6 - 7 | 824 | 40044874 | 90 | |
BC103555.1 | - | 6 - 7 | 824 | 40044870 | 90 | |
BC103556.1 | - | 6 - 7 | 824 | 40044871 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
intrinsic apoptotic signaling pathway in response to DNA damage
cell differentiation
extrinsic apoptotic signaling pathway in absence of ligand
cysteine-type endopeptidase activity
peptidase activity
cysteine-type peptidase activity
hydrolase activity
cysteine-type endopeptidase activity involved in apoptotic signaling pathway
proteolysis
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
intrinsic apoptotic signaling pathway in response to DNA damage
cell differentiation
extrinsic apoptotic signaling pathway in absence of ligand
cysteine-type endopeptidase activity
peptidase activity
cysteine-type peptidase activity
hydrolase activity
cysteine-type endopeptidase activity involved in apoptotic signaling pathway
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064063 | AJ007750.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064063 | AK081645.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064063 | AF092997.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064063 | NM_009809.5 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 518443 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 343058 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 887657 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 772262 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165771 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106342 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137474 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139077 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064063 | AJ007750.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064063 | AK081645.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064063 | AF092997.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064063 | NM_009809.5 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P463159 | AJ007750.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P463159 | AK081645.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P463159 | AF092997.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P463159 | NM_009809.5 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275364 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275377 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275357 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275366 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275373 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275352 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275361 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275368 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275375 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275342 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275348 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275359 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275344 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275355 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275360 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275363 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275365 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275346 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275350 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275347 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27275370 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137474 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106342 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139077 | - | - |
Whole Mouse Genome G4122A | A_51_P463159 | AJ007750.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463159 | AK081645.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463159 | AF092997.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463159 | NM_009809.5 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129930 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137474 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106342 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139077 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08757878 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08757884 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08261254 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08757882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27275361 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275359 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275334 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275341 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275293 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08387080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08563098 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275346 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275382 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275319 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275361 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275352 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08557964 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275326 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275327 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275313 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08757878 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08429546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275328 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275322 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275336 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08252081 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275388 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275368 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275296 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08761894 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08252079 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08488087 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275355 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275329 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275371 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275315 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275383 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275290 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275373 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275335 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275350 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275380 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08429550 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275344 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275312 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275349 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08252080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08261253 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275366 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275339 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275372 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275318 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275291 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08261254 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275370 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275292 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27275320 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10370526 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10370523 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10370527 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10370524 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10370529 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10370528 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10370525 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1443787_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418748_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10370522 | - | - |
Mouse Genome 430 2.0 Array | 1443787_x_at | - | - |
Mouse Genome 430 2.0 Array | 1443787_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1418748_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418748_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12365_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12365_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1000017927.mm.1 | NM_009809.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000017927.mm.1 | NM_009809.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000017927.mm.1 | TC1000002460.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000017928.mm.1 | NM_009809.5.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000017928.mm.1 | NM_009809.5 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000017928.mm.1 | TC1000002460.mm.1 | MTA TC |
Mouse Expression Set 430 | 1418748_PM_at | - | - |
Mouse Expression Set 430 | 1418748_at | - | - |
Mouse Expression Set 430 | 1443787_PM_x_at | - | - |
Mouse Expression Set 430 | 1443787_x_at | - | - |
Murine Genome U74v2 Set | 103837_PM_at | - | - |
Murine Genome U74v2 Set | 103837_at | - | - |
Mouse Genome 430A 2.0 Array | 1418748_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418748_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418748_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418748_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443787_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443787_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 455803 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6510156 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006510156 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1241322 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1241322 | - | - |
mouse-6 v1.1 (Illumina) | scl37766.7.1_51-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6510156 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl37766.7.1_51-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6510156 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1241322 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1241322 | - | - |
