| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900004497.mm.1 |
NM_001104616.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004497.mm.1 |
NM_001104616.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004497.mm.1 |
NM_001104617.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004497.mm.1 |
NM_001104617.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004497.mm.1 |
NM_009041.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004497.mm.1 |
NM_009041.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900004497.mm.1 |
TC0900000626.mm.1 |
MTA TC |