Assay Details
Gene Symbol: 3110039I08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014147.1 | - | 114 | 63 | ||
AW491891.1 | - | 63 | ||||
BB625832.1 | - | 148 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1700063D05 gene | 1700063D05Rik | -158.8kb | Sense | Chr. 9 - 41206155 - 41217627 on NCBI Build GRCm38 | |
Downstream | microRNA 100 | Mir100 |
Mirn100, mir-100, mmu-mir-100 |
138.1kb | Sense | Chr. 9 - 41531425 - 41531504 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
