Assay Details
Gene Symbol: Crtam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF001104.1 | - | 1 - 2 | 44 | 66 | |
BC117715.1 | - | 1 - 2 | 44 | 66 | ||
BC117716.1 | - | 1 - 2 | 44 | 40088099 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
