Assay Details
Gene Symbol: Crtam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281954.1 | NP_001268883.1 | 3 - 4 | 379 | 76 | |
NM_019465.4 | NP_062338.3 | 3 - 4 | 379 | 76 | ||
GenBank mRNA | AF001104.1 | - | 3 - 4 | 337 | 76 | |
AK005719.1 | - | 1 - 2 | 102 | 76 | ||
BC117715.1 | - | 3 - 4 | 337 | 76 | ||
BC117716.1 | - | 3 - 4 | 337 | 40088099 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK005719.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
