Assay Details
| Gene Symbol: Npat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081152.1 | NP_001074621.1 | 12 - 13 | 1238 | 132 | |
| GenBank mRNA | AK033012.1 | - | 12 - 13 | 1218 | 132 | |
| AK080294.1 | - | 12 - 13 | 1239 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npat
Genomic Map