Assay Details
| Gene Symbol: Ddx10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029936.2 | NP_084212.2 | 12 - 13 | 1608 | 115 | |
| GenBank mRNA | AK019495.1 | - | 12 - 13 | 1588 | 115 | |
| AK052540.1 | - | 12 - 13 | 1588 | 115 | ||
| BC023303.1 | - | 12 - 13 | 1556 | 115 | ||
| BC049261.1 | - | 12 - 13 | 1549 | 115 | ||
| BC055481.1 | - | 11 - 12 | 1326 | 115 | ||
| BC080729.1 | - | 12 - 13 | 1576 | 6504816 | 115 | |
More Information
Set Membership: |
|

Ddx10
Genomic Map