Assay Details
| Gene Symbol: Kdelc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212445.2 | NP_997610.1 | 2 - 3 | 459 | 86 | |
| GenBank mRNA | AK075811.1 | - | 2 - 3 | 268 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdelc2
Genomic Map