Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000005023.mm.1 |
NM_001145826.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005023.mm.1 |
NM_001145826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005023.mm.1 |
TC1000000710.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000005020.mm.1 |
NM_001145826.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005020.mm.1 |
NM_001145826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005020.mm.1 |
TC1000000710.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000005027.mm.1 |
NM_001145826.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005027.mm.1 |
NM_001145826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005027.mm.1 |
NM_153406.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005027.mm.1 |
NM_153406.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005027.mm.1 |
TC1000000710.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000005021.mm.1 |
NM_001145826.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005021.mm.1 |
NM_001145826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005021.mm.1 |
TC1000000710.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000005022.mm.1 |
NM_001145826.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005022.mm.1 |
NM_001145826.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005022.mm.1 |
TC1000000710.mm.1 |
MTA TC |